missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorophenoxyacetic acid, 98+%, Thermo Scientific™

Product Code. 11419747
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419747 5 g 5g
11409747 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419747 Supplier Thermo Scientific Alfa Aesar Supplier No. B22731.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14892-14-9
Molecular Formula C8H3F5O3
Molecular Weight (g/mol) 242.101
MDL Number MFCD00004297
InChI Key SMXPFEBIAASLOR-UHFFFAOYSA-N
Synonym pentafluorophenoxyacetic acid, pentafluorophenoxy acetic acid, 2,3,4,5,6-pentafluorophenoxyacetic acid, pentafluorophenyloxyacetic acid, acetic acid, pentafluorophenoxy, 2,3,4,5,6-pentafluorophenoxy acetic acid, 2-2,3,4,5,6-pentafluorophenoxy acetic acid, pentafluorophenyloxy-acetic acid
PubChem CID 84680
IUPAC Name 2-(2,3,4,5,6-pentafluorophenoxy)acetic acid
SMILES C(C(=O)O)OC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point 108°C to 110°C
Quantity 5 g
Beilstein 1888139
Formula Weight 242.1
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorophenoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.