missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorophenylacetic acid, 98+%

Product Code. 11480567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480567 1 g 1g
11490567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480567 Supplier Thermo Scientific Alfa Aesar Supplier No. B22430.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 653-21-4 | C8H3F5O2 | 226.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-21-4
Molecular Formula C8H3F5O2
Molecular Weight (g/mol) 226.102
MDL Number MFCD00004316
InChI Key LGCODSNZJOVMHV-UHFFFAOYSA-N
Synonym pentafluorophenylacetic acid, 2,3,4,5,6-pentafluorophenylacetic acid, pentafluorophenyl acetic acid, 2-perfluorophenyl acetic acid, 2-2,3,4,5,6-pentafluorophenyl acetic acid, 2,3,4,5,6-pentafluorphenylacetic acid, benzeneacetic acid, 2,3,4,5,6-pentafluoro, acmc-20aoj2, pentafluorophenylaceticacid, 2-pentafluorophenyl acetic acid
PubChem CID 69553
IUPAC Name 2-(2,3,4,5,6-pentafluorophenyl)acetic acid
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)C(=O)O

Specifications

Melting Point 107°C to 110°C
Quantity 1 g
Beilstein 2056612
Formula Weight 226.1
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.