missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrachlorotoluene, TRC

Product Code. 17994175
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994175 10 mg 10mg
17974225 50 mg 50mg
17901605 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994175 Supplier Toronto Research Chemicals Supplier No. TRCT29205010MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1006-32-2
Molecular Formula C7H4Cl4
Molecular Weight (g/mol) 229.92
Synonym Toluene, 2,3,4,5-tetrachloro- (6CI,7CI,8CI), 1,2,3,4-Tetrachloro-5-methylbenzene, 2,3,4,5-Tetrachlorotoluene
IUPAC Name 1,2,3,4-tetrachloro-5-methylbenzene
SMILES Cc1cc(Cl)c(Cl)c(Cl)c1Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H4Cl4/c1-3-2-4(8)6(10)7(11)5(3)9/h2H,1H3
Formula Weight 227.9067
Percent Purity >95%
Chemical Name or Material 2,3,4,5-Tetrachlorotoluene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.