missing translation for 'onlineSavingsMsg'
Learn More

2-[3,4-(Methylenedioxy)phenyl]ethylamine hydrochloride, 95%

Product Code. 15465556
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465556 5 g 5g
15455556 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465556 Supplier Thermo Scientific Alfa Aesar Supplier No. H60208.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1653-64-1 | C9H12ClNO2 | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1653-64-1
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00035193
InChI Key NDYXFQODWGEGNU-UHFFFAOYSA-N
Synonym 3,4-methylenedioxyphenethylamine hydrochloride, 2-3,4-methylenedioxyphenyl ethylamine hydrochloride, phenethylamine, 3,4-methylenedioxy-, hydrochloride, 2-benzo d 1,3 dioxol-5-yl ethanamine hydrochloride, homopiperonylamine hydrochloride, 1,3-benzodioxole-5-ethanamine, hydrochloride, 3,4-methylenedioxy-beta-phenylethylamine hydrochloride, methylenedioxyphenethylamine hydrochloride, 3,4-methylenedioxy phenylethylamine hydrochloride, 3,4-methylenedioxy-.beta.-phenylethylamine hydrochloride
PubChem CID 74246
IUPAC Name 2-(1,3-benzodioxol-5-yl)ethanamine;hydrochloride
SMILES C1OC2=C(O1)C=C(C=C2)CCN.Cl

Specifications

Melting Point 213°C to 214°C
Quantity 5 g
Formula Weight 201.65
Percent Purity 95%
Chemical Name or Material 2-[3,4-(Methylenedioxy)phenyl]ethylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.