missing translation for 'onlineSavingsMsg'
Learn More

2-[3,5-Dichloro-4-(4-chlorobenzoyl)phenyl]-1,2,4-triazine-3,5(2H,4H)-dione, TRC

Product Code. 30005148
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005148 10 mg 10mg
30008463 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005148 Supplier Toronto Research Chemicals Supplier No. TRCD43400510MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133648-81-4
Molecular Formula C16 H8 Cl3 N3 O3
Molecular Weight (g/mol) 396.61
Synonym 1,2,4-Triazine-3,5(2H,4H)-dione, 2-[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]-, 2-[3,5-Dichloro-4-(4-chlorobenzoyl)phenyl]-1,2,4-triazine-3,5(2H,4H)-dione, Diclazuril Ketone
IUPAC Name 2-[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]-1,2,4-triazine-3,5-dione
SMILES Clc1ccc(cc1)C(=O)c2c(Cl)cc(cc2Cl)N3N=CC(=O)NC3=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H8Cl3N3O3/c17-9-3-1-8(2-4-9)15(24)14-11(18)5-10(6-12(14)19)22-16(25)21-13(23)7-20-22/h1-7H,(H,21,23,25)
Formula Weight 394.9631
Percent Purity >95%
Chemical Name or Material 2-[3,5-Dichloro-4-(4-chlorobenzoyl)phenyl]-1,2,4-triazine-3,5(2H,4H)-dione
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.