missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylphenol, TRC

Product Code. 30455652
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455652 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455652 Supplier Toronto Research Chemicals Supplier No. TRCT796595100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,5-Trimethylphenol
CAS 697-82-5
Molecular Formula C9 H12 O
Quantity 100 g
Synonym 2,3,5-Trimethylphenol, Phenol','2,3,5-trimethyl- (6CI','8CI','9CI','ACI), 2,3,5-Trimethylphenol (ACI), 1-Hydroxy-2,3,5-trimethylbenzene, 3,5,6-Trimethylphenol, 6-Hydroxypseudocumene, Isopseudocumenol, NSC 5296
InChI Formula InChI=1S/C9H12O/c1-6-4-7(2)8(3)9(10)5-6/h4-5,10H,1-3H3
SMILES Cc1cc(C)c(C)c(O)c1
IUPAC Name 2,3,5-trimethylphenol
Molecular Weight (g/mol) 136.19
Formula Weight 136.0888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.