missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trifluorobenzaldehyde, 97%, Thermo Scientific™

Product Code. 11469836
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11469836

Brand: Thermo Scientific Alfa Aesar B20319.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104451-70-9
Molecular Formula C7H3F3O
Molecular Weight (g/mol) 160.10
MDL Number MFCD00061195
InChI Key XSBAHBVACIKRTG-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3,6-trifluoro, 2,3,6-trifluoro-benzaldehyde, pubchem1467, acmc-1bqm6, 2,3,6-trifluorobenzoaldehyde, ksc494g2t, 2,3,6-trifluoro benzaldehyde, timtec-bb sbb003970, attercop-chm at111497
PubChem CID 517845
IUPAC Name 2,3,6-trifluorobenzaldehyde
SMILES FC1=CC=C(F)C(C=O)=C1F

Specifications

Density 1.425
Boiling Point 173°C
Flash Point 81°C (177°F)
Refractive Index 1.488
Quantity 5 g
Beilstein 7701780
Sensitivity Air Sensitive
Formula Weight 160.1
Percent Purity 97%
Chemical Name or Material 2,3,6-Trifluorobenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.