Learn More
2,3-Bis[(acetyloxy)methyl]-1-oxoquinoxalin-1-ium-4(1H)-olate, TRC
Product Code. 30010416
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010416 10 mg 10mg
30004118 50 mg 50mg
30010393 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010416 Supplier Toronto Research Chemicals Supplier No. TRCB32562010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10103-89-6
Molecular Formula C14 H14 N2 O6
Molecular Weight (g/mol) 306.27
Synonym 2,3-Di(acetoxymethyl)quinoxaline di-N-Oxide;
IUPAC Name [3-(acetoxymethyl)-1,4-dioxo-quinoxaline-1,4-diium-2-yl]methyl acetate
SMILES CC(=O)OCC1=C(COC(=O)C)[N+](=O)c2ccccc2[N+]1=O
Specifications
Analyte or Component Names 2,3-Bis[(acetyloxy)methyl]-1-oxoquinoxalin-1-ium-4(1H)-olate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H14N2O6/c1-9(17)21-7-13-14(8-22-10(2)18)16(20)12-6-4-3-5-11(12)15(13)19/h3-6H,7-8H2,1-2H3/q+2
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Formula Weight 306.0841
Chemical Name or Material 2,3-Bis[(acetyloxy)methyl]-1-oxoquinoxalin-1-ium-4(1H)-olate