missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5-(trifluoromethyl)pyridine, 97%

Product Code. 10503791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10503791 25 g Glass bottle 25g
10474155 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10503791 Supplier Thermo Scientific Acros Supplier No. 241810250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69045-84-7 | C6H2Cl2F3N | 215.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69045-84-7
Molecular Formula C6H2Cl2F3N
Molecular Weight (g/mol) 215.98
MDL Number MFCD00042243
InChI Key ABNQGNFVSFKJGI-UHFFFAOYSA-N
Synonym 2,3-dichloro-5-trifluoromethyl pyridine, unii-5f2c8lx8ie, 2,3-dichloro-5-trifluoro methylpyridine, 2,3,5-dctf, 5f2c8lx8ie, 2,3-dichloro-5-trifluoromethyl-pyridine, pyridine, 2,3-dichloro-5-trifluoromethyl, pubchem9237
PubChem CID 112234
IUPAC Name 2,3-dichloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CN=C(Cl)C(Cl)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 8.0°C to 9.0°C
Color Colorless to Yellow
Density 1.5490g/mL
Boiling Point 176.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4745 to 1.4765
Quantity 25 g
Specific Gravity 1.549
Formula Weight 216
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Dichloro-5-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Dichloro-5-(trifluoromethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.