missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorobenzaldehyde, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2,3-Dichlorobenzaldehyde |
| CAS | 6334-18-5 |
| Molecular Formula | C7 H4 Cl2 O |
| Quantity | 1 g |
| Synonym | Benzaldehyde, 2,3-dichloro-, 2,3-Dichlorobenzaldehyde, NSC 28450 |
| InChI Formula | InChI=1S/C7H4Cl2O/c8-6-3-1-2-5(4-10)7(6)9/h1-4H |
| SMILES | Clc1cccc(C=O)c1Cl |
| IUPAC Name | 2,3-dichlorobenzaldehyde |
| Molecular Weight (g/mol) | 175.01 |
| Formula Weight | 173.9639 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction