missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoyl chloride, TRC

Product Code. 30373898
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373898 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373898 Supplier Toronto Research Chemicals Supplier No. TRCD4317055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dichlorobenzoyl chloride
CAS 2905-60-4
Molecular Formula C7 H3 Cl3 O
Quantity 5 g
Synonym 2,3-Dichlorobenzoyl chloride
InChI Formula InChI=1S/C7H3Cl3O/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3H
SMILES ClC(=O)c1cccc(Cl)c1Cl
IUPAC Name 2,3-dichlorobenzoyl chloride
Molecular Weight (g/mol) 209.457
Formula Weight 207.925
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.