missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-5-(trifluoromethyl)pyridine, 98%

Product Code. 11445740
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445740 1 g 1g
11455740 5 g 5g
11465740 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445740 Supplier Thermo Scientific Alfa Aesar Supplier No. H25958.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89402-42-6 | C6H2F5N | 183.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89402-42-6
Molecular Formula C6H2F5N
Molecular Weight (g/mol) 183.08
MDL Number MFCD04972837
InChI Key XIFCGIKPAAZFFS-UHFFFAOYSA-N
Synonym 2,3-difluoro-5-trifluoromethyl pyridine, 2,3-difluoro-5-trifluoromethyl-pyridine, pyridine,2,3-difluoro-5-trifluoromethyl, pyridine, 2,3-difluoro-5-trifluoromethyl, acmc-20dwi8, 2,3-di-fluoro-5-trifluoromethyl pyridine, alpha,alpha,alpha,5,6-pentafluoro-3-picoline, 2,3-bis fluoranyl-5-trifluoromethyl pyridine
PubChem CID 13421547
IUPAC Name 2,3-difluoro-5-(trifluoromethyl)pyridine
SMILES FC1=C(F)N=CC(=C1)C(F)(F)F

Specifications

Density 1.47
Boiling Point 104°C
Flash Point >100°C (212°F)
Quantity 1 g
Formula Weight 183.08
Percent Purity 98%
Chemical Name or Material 2,3-Difluoro-5-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.