Learn More
2,3-Dihydro-1H-indene-2-carboxylic acid, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 25177-85-9 |
| Molecular Formula | C10H10O2 |
| MDL Number | MFCD00085095 |
| Quantity | 10 g |
| SMILES | OC(=O)C1CC2=CC=CC=C2C1 |
| Molecular Weight (g/mol) | 162.19 |
| Percent Purity | 95% |
| Chemical Name or Material | 2,3-Dihydro-1H-indene-2-carboxylic acid |