missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylic Acid tert-Butyl Ester, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 2,3-Dihydro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylic Acid tert-Butyl Ester |
| CAS | 837392-67-3 |
| Molecular Formula | C19H28BNO4 |
| Quantity | 250 mg |
| Synonym | tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline-1-carboxylate 2,3-Dihydro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylic Acid 1,1-Dimethylethyl Ester |
| InChI Formula | InChI=1S/C19H28BNO4/c1-17(2,3)23-16(22)21-11-10-13-12-14(8-9-15(13)21)20-24-18(4,5)19(6,7)25-20/h8-9,12H,10-11H2,1-7H3 |
| SMILES | CC(C)(C)OC(=O)N1CCc2cc(ccc12)B3OC(C)(C)C(C)(C)O3 |
| Molecular Weight (g/mol) | 345.24 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction