missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-5,6-dimethoxy-2-[(1-oxido-4-pyridinyl)methylene]-1H-inden-1-one, TRC

Product Code. 30010028
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010028 250 mg 250mg
30004811 10 mg 10mg
30007153 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010028 Supplier Toronto Research Chemicals Supplier No. TRCD450165250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 896134-06-8
Molecular Formula C17 H15 N O4
Molecular Weight (g/mol) 297.3
Synonym 2,3-Dihydro-5,6-dimethoxy-2-[(1-oxido-4-pyridinyl)methylene]-1H-inden-1-one;
IUPAC Name 5,6-dimethoxy-2-[(1-oxidopyridin-1-ium-4-yl)methylene]indan-1-one
SMILES COc1cc2CC(=Cc3cc[n+]([O-])cc3)C(=O)c2cc1OC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H15NO4/c1-21-15-9-12-8-13(7-11-3-5-18(20)6-4-11)17(19)14(12)10-16(15)22-2/h3-7,9-10H,8H2,1-2H3
Formula Weight 297.1001
Chemical Name or Material 4-(5,6-Dimethoxy-1-oxoindan-2-ylidene)methylpyridine 1-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.