missing translation for 'onlineSavingsMsg'
Learn More

2,3-dihydroxypropyl 4-hydroxybenzoate, TRC

Product Code. 30137132
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137132 5 mg 5mg
30137924 50 mg 50mg
17974245 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137132 Supplier Toronto Research Chemicals Supplier No. TRCD4585555MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93778-15-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
SMILES C1=CC(=CC=C1C(=O)OCC(CO)O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H12O5/c11-5-9(13)6-15-10(14)7-1-3-8(12)4-2-7/h1-4,9,11-13H,5-6H2
Formula Weight 212.0685
Chemical Name or Material 2,3-dihydroxypropyl 4-hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.