missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethoxy-5-methyl-p-benzoquinone, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2,3-Dimethoxy-5-Methylbenzoquinone |
| CAS | 605-94-7 |
| Molecular Formula | C9 H10 O4 |
| Quantity | 1 g |
| Synonym | CoQ0, Ubiquinone Q0 |
| InChI Formula | InChI=1S/C9H10O4/c1-5-4-6(10)8(12-2)9(13-3)7(5)11/h4H,1-3H3 |
| SMILES | COC1=C(OC)C(=O)C(=CC1=O)C |
| IUPAC Name | 2,3-dimethoxy-5-methyl-1,4-benzoquinone |
| Molecular Weight (g/mol) | 182.17 |
| Formula Weight | 182.0579 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction