missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Hydroxymethyl)-5-methyl-4-triazolyl]-5-chlorobenzophenone, TRC

Product Code. 17950575
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17950575 5 mg 5mg
17924935 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17950575 Supplier Toronto Research Chemicals Supplier No. TRCH9479505MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38150-27-5
Molecular Formula C17 H14 Cl N3 O2
Molecular Weight (g/mol) 327.76
Synonym Alprazolam Imp. B (EP), [5-Chloro-2-(3-hydroxymehtyl-5-methyl-4H-1,2,4-triazol-4-yl)phenyl]phenylmethanone, 2-[3-(Hydroxymethyl)-5-methyl-4-triazolyl]-5-chlorobenzophenone
IUPAC Name [5-chloro-2-[3-(hydroxymethyl)-5-methyl-1,2,4-triazol-4-yl]phenyl]-phenylmethanone
SMILES Cc1nnc(CO)n1c2ccc(Cl)cc2C(=O)c3ccccc3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H14ClN3O2/c1-11-19-20-16(10-22)21(11)15-8-7-13(18)9-14(15)17(23)12-5-3-2-4-6-12/h2-9,22H,10H2,1H3
Formula Weight 327.0775
Chemical Name or Material 2-[3-(Hydroxymethyl)-5-methyl-4-triazolyl]-5-chlorobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.