missing translation for 'onlineSavingsMsg'
Learn More

2,3-Lutidine, 99%

Product Code. 11372266
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372266 25 g 25g
11382266 100 g 100g
11392266 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11372266 Supplier Thermo Scientific Alfa Aesar Supplier No. L03697.14
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 583-61-9 | C7H9N | 107.156 g/mol

2,3-Lutidine is a lansoprazole intermediates. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Lutidine is a lansoprazole intermediates. It is used as pharmaceutical intermediate.

Solubility
Soluble in water 95 gl/ @ 20˚C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-61-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00009605
InChI Key HPYNZHMRTTWQTB-UHFFFAOYSA-N
Synonym 2,3-lutidine, pyridine, 2,3-dimethyl, pyridine, dimethyl, dimethylpyridine, unii-5q0f649h6g, 2,3-dimethyl-pyridine, 2-3-lutidine, pyridine,3-dimethyl, 2,3-d1methylpyridine, pubchem20520
PubChem CID 11420
IUPAC Name 2,3-dimethylpyridine
SMILES CC1=C(N=CC=C1)C

Specifications

Melting Point -17°C
Density 0.948
Boiling Point 160°C to 161°C
Flash Point 50°C (122°F)
Odor Unpleasant
Refractive Index 1.508
Quantity 25 g
UN Number UN2929
Beilstein 106418
Solubility Information Soluble in water 95 gl/ @ 20?C.
Formula Weight 107.16
Percent Purity 99%
Chemical Name or Material 2,3-Lutidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.