missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetrafluoroisophthalonitrile, TRC

Product Code. 30409036
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409036 1 g 1g
30421686 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30409036 Supplier Toronto Research Chemicals Supplier No. TRCT2929151G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,5,6-Tetrafluoroisophthalonitrile
CAS 2377-81-3
Molecular Formula C8F4N2
Quantity 1 g
Synonym Tetrafluoroisophthalonitrile, Perfluoroisophthalonitrile
InChI Formula InChI=1S/C8F4N2/c9-5-3(1-13)6(10)8(12)7(11)4(5)2-14
SMILES Fc1c(F)c(C#N)c(F)c(C#N)c1F
IUPAC Name 2,4,5,6-tetrafluorobenzene-1,3-dicarbonitrile
Molecular Weight (g/mol) 200.09
Formula Weight 199.9998
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.