missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorophenol, 98%, ACROS Organics™

Product Code. 10760781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10760781 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10760781 Supplier Thermo Scientific Acros Supplier No. 308680010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2268-16-8
CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 42°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H3F3O
Linear Formula F3C6HOH
MDL Number MFCD00061215
Quantity 1 g
Synonym phenol, 2,4,5-trifluoro, phenol,2,4,5-trifluoro, 2,4,5-tfifluorophenol, 2,4,5-trifluoro-phenol, 2,4,5-?trifluorophenol, acmc-1cne2, 2,4,5-trifluorophenol, 2,4,5-tris fluoranyl phenol, 2,4,5-trifluorophenol 1gr
InChI Key ODGMYCITQAIRCI-UHFFFAOYSA-N
SMILES C1=C(C(=CC(=C1F)F)F)O
IUPAC Name 2,4,5-trifluorophenol
Molecular Weight (g/mol) 148.084
PubChem CID 123153
Formula Weight 148.09
Percent Purity ≥97.5%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2, 4, 5-Trifluorophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.