missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethoxybenzaldehyde, 98%

Product Code. 11469827
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469827 25 g 25g
11479827 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469827 Supplier Thermo Scientific Alfa Aesar Supplier No. B22792.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4460-86-0 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4460-86-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00003312
InChI Key IAJBQAYHSQIQRE-UHFFFAOYSA-N
Synonym asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296
PubChem CID 20525
IUPAC Name 2,4,5-trimethoxybenzaldehyde
SMILES COC1=CC(OC)=C(C=O)C=C1OC

Specifications

Melting Point 111°C to 115°C
Boiling Point 140°C (4 mmHg)(sublimation)
Quantity 25 g
Beilstein 1951403
Sensitivity Air Sensitive
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 2,4,5-Trimethoxybenzaldehyde
compliance-icons
Product Identifier
  • 2,4,5-Trimethoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,4,5-Trimethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.