missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethoxybenzaldehyde, TRC

Product Code. 30449575
Change view
Click to view available options
Quantity:
10 mg
50 mg
25 g
Unit Size:
10mg
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449575 25 g 25g
30452503 10 mg 10mg
30450584 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449575 Supplier Toronto Research Chemicals Supplier No. TRCT79574025G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4460-86-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.2
Synonym 2,4,5-Trimethoxybenzaldehyde, 2,4,5-Trimethoxylbenzaldehyde, 3,4,6-Trimethoxybenzaldehyde, Asaraldehyde, Asaronaldehyde, Asarylaldehyde, Gazarin, NSC 89299
IUPAC Name 2,4,5-trimethoxybenzaldehyde
SMILES COc1cc(OC)c(C=O)cc1OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H12O4/c1-12-8-5-10(14-3)9(13-2)4-7(8)6-11/h4-6H,1-3H3
Formula Weight 196.0736
Chemical Name or Material 2,4,5-Trimethoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.