missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethylaniline Hydrochloride, TRC

Product Code. 18083050
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18083050 500 mg 500mg
18043550 250 mg 250mg
18001160 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18083050 Supplier Toronto Research Chemicals Supplier No. TRCT214360500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21436-97-5
Molecular Formula C9 H13 N . Cl H
Molecular Weight (g/mol) 171.67
Synonym 2,4,5-Trimethylaniline Hydrochloride, Aniline, 2,4,5-trimethyl-, hydrochloride (8CI), Benzenamine, 2,4,5-trimethyl-, hydrochloride (9CI), 2,4,5-Trimethylaniline hydrochloride, Pseudocumidine hydrochloride
IUPAC Name 2,4,5-trimethylaniline;hydrochloride
SMILES Cl.Cc1cc(C)c(N)cc1C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H13N.ClH/c1-6-4-8(3)9(10)5-7(6)2;/h4-5H,10H2,1-3H3;1H
Formula Weight 171.0815
Percent Purity >95%
Chemical Name or Material 2,4,5-Trimethylaniline Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.