Learn More
2,4,6-Triallyloxy-1,3,5-triazine, TRC
Product Code. 30455511
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455511 500 mg 500mg
30449179 2.5 g 2.5g
30455211 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30455511 Supplier Toronto Research Chemicals Supplier No. TRCT767158500MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101-37-1
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
IUPAC Name 2,4,6-tris(prop-2-enoxy)-1,3,5-triazine
SMILES C=CCOc1nc(OCC=C)nc(OCC=C)n1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C12H15N3O3/c1-4-7-16-10-13-11(17-8-5-2)15-12(14-10)18-9-6-3/h4-6H,1-3,7-9H2
Formula Weight 249.1113
Chemical Name or Material 2,4,6-Triallyloxy-1,3,5-triazine