missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluoroaniline, 97%, ACROS Organics™

Product Code. 10052570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10052570 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10052570 Supplier Thermo Scientific Acros Supplier No. 188410010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 363-81-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 33°C to 37°C
Color Colorless to Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H4F3N
Linear Formula F3C6H2NH2
MDL Number MFCD00007650
Quantity 1 g
Synonym 2-amino-1,3,5-trifluorobenzene, 2,4,6-trifluorobenzenamine, benzenamine, 2,4,6-trifluoro, 2,4,6-trifluorophenylamine, pubchem2468, acmc-1cmhu, 2,4,6 trifluoroaniline, 2,4,6-tri-fluoroaniline, 2,4,6-trifluoro aniline, 2,4,6-trifluoro-aniline
InChI Key BJSVKBGQDHUBHZ-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1F)N)F)F
IUPAC Name 2,4,6-trifluoroaniline
Molecular Weight (g/mol) 147.1
PubChem CID 67765
Formula Weight 147.1
Percent Purity ≥96.0%
Physical Form Crystals
Chemical Name or Material 2, 4, 6-Trifluoroaniline, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.