missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylphenol-d11, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | 2,4,6-Trimethylphenol-d11 |
| CAS | 362049-45-4 |
| Molecular Formula | C9 D11 H O |
| Quantity | 1 g |
| Synonym | Phen-3,5-d2-ol, 2,4,6-tri(methyl-d3)- (9 CI) |
| InChI Formula | InChI=1 S/C9H12O/c1-6-4-7(2)9(10)8(3)5-6/h4-5,10 H,1-3H3/i1D3,2D3,3D3,4 D,5 D |
| SMILES | [2 H]c1c(c([2 H])c(c(O)c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 3,5-dideuterio-2,4,6-tris(trideuteriomethyl)phenol |
| Molecular Weight (g/mol) | 147.26 |
| Formula Weight | 147.1579 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction