missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(dimethylaminomethyl)phenol, Thermo Scientific™

Product Code. 17730569 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17730569 25 g 25g
17740569 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17730569 Supplier Thermo Scientific Acros Supplier No. 469290250
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

Alpha Vector TRISDIMETHYLAMINOMETHYLPHENOL
Chemical Name or Material 2,4,6-Tris(dimethylaminomethyl)phenol
CAS 90-72-2
Color Colorless to Yellow
Flash Point 149°C
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C15 H27 N3 O
Refractive Index 1.5150 to 1.5170 (20°C, 589 nm)
MDL Number 00008330
Quantity 25 g
UN Number 2735
Molecular Weight (g/mol) 265.4
Physical Form Viscous Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.