missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Boc-piperazino)pyrimidine-5-boronic acid pinacol ester, TRC

Product Code. 30335966
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335966 250 mg 250mg
30346782 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30335966 Supplier Toronto Research Chemicals Supplier No. TRCB663205250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(4-Boc-piperazino)pyrimidine-5-boronic acid pinacol ester
CAS 940284-98-0
Molecular Formula C19H31BN4O4
Quantity 250 mg
InChI Formula InChI=1S/C19H31BN4O4/c1-17(2,3)26-16(25)24-10-8-23(9-11-24)15-21-12-14(13-22-15)20-27-18(4,5)19(6,7)28-20/h12-13H,8-11H2,1-7H3
SMILES CC(C)(C)OC(=O)N1CCN(CC1)c2ncc(cn2)B3OC(C)(C)C(C)(C)O3
IUPAC Name tert-butyl 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl]piperazine-1-carboxylate
Molecular Weight (g/mol) 390.29
Formula Weight 390.2438
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.