Learn More
2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol, Dr. Ehrenstorfer
Product Code. 30326211
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326211 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326211 Supplier Dr. Ehrenstorfer Supplier No. DREC10931123
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol
CAS 3864-99-1
Analyte or Component Names 2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol
Shipping Condition Room Temperature
Molecular Formula C20 H24 Cl N3 O
Certifications/Compliance ISO 17034
Synonym Phenol, 2,4-di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)- (7CI,8CI), 2-(5-Chloro-2H-benzotriazol-2-yl)-4,6-bis(1,1-dimethylethyl)phenol, 2,4-Di-(tert-butyl)-6-(5-chloro-2H-benzo[d][1,2,3]triazol-2-yl)phenol, 2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol, 2,4-Di-tert-butyl-6-(5-chlorobenzotriazol-2-yl)phenol, 2-(2-Hydroxy-3,5-di-tert-butylphenyl)-5-chloro-2H-benzotriazole, 2-(2-Hydroxy-3,5-di-tert-butylphenyl)-5-chlorobenzotriazole, 2-(2'-Hydroxy-3',5'-di-tert-butylphenyl)-5-chlorobenzotriazole, 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-5-chloro-2H-benzotriazole, 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-5-chlorobenzotriazole, 2-(3',5'-Di-tert-butyl-2'-hydroxyphenyl)-5-chlorobenzotriazole, 5-Chloro-2-(2-hydroxy-3,5-di-tert-butylphenyl)-2H-benzotriazole, 5-Chloro-2-(2-hydroxy-3,5-di-tert-butylphenyl)benzotriazole, 5-Chloro-2-(3,5-di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole, 5-Chloro-2-(3,5-di-tert-butyl-2-hydroxyphenyl)benzotriazole, 5-Chloro-2-(3',5'-di-tert-butyl-2'-hydroxyphenyl)benzotriazole, ADK Stab LA 34, Antioxidant 327, Chiguard 327, Cyasorb UV 5357, Eversorb 75, Hisorb 327, Hisorp 327, Kemisorb 72, LA 34, Light stabilizer 327, Lowilite 27, Mark LA 34, Seesorb 702, Songsorb 3270, TNV 327, Tinuvin 327, UV 2, UV 2 (UV stabilizer), UV 327, UV-Chek AM 607, UVP 327, Viosorb 580
InChI Formula InChI=1S/C20H24ClN3O/c1-19(2,3)12-9-14(20(4,5)6)18(25)17(10-12)24-22-15-8-7-13(21)11-16(15)23-24/h7-11,25H,1-6H3
Solution Type Neat
SMILES CC(C)(C)c1cc(c(O)c(c1)C(C)(C)C)n2nc3ccc(Cl)cc3n2
IUPAC Name 2,4-ditert-butyl-6-(5-chlorobenzotriazol-2-yl)phenol
Industry Type Environmental Testing
Molecular Weight (g/mol) 357.88
Quantity 50 mg
Formula Weight 357.1608 g/mol