Learn More
2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol, TRC
Product Code. 30384260
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384260 10 g
30370304 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30384260 Supplier Toronto Research Chemicals Supplier No. TRCD42801510G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3864-99-1
Molecular Formula C20 H24 Cl N3 O
Molecular Weight (g/mol) 357.88
Synonym Phenol, 2,4-di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)- (7CI,8CI), 2-(5-Chloro-2H-benzotriazol-2-yl)-4,6-bis(1,1-dimethylethyl)phenol, 2,4-Di-(tert-butyl)-6-(5-chloro-2H-benzo[d][1,2,3]triazol-2-yl)phenol, 2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol, 2,4-Di-tert-butyl-6-(5-chlorobenzotriazol-2-yl)phenol, 2-(2-Hydroxy-3,5-di-tert-butylphenyl)-5-chloro-2H-benzotriazole, 2-(2-Hydroxy-3,5-di-tert-butylphenyl)-5-chlorobenzotriazole, 2-(2'-Hydroxy-3',5'-di-tert-butylphenyl)-5-chlorobenzotriazole, 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-5-chloro-2H-benzotriazole, 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-5-chlorobenzotriazole, 2-(3',5'-Di-tert-butyl-2'-hydroxyphenyl)-5-chlorobenzotriazole, 5-Chloro-2-(2-hydroxy-3,5-di-tert-butylphenyl)-2H-benzotriazole, 5-Chloro-2-(2-hydroxy-3,5-di-tert-butylphenyl)benzotriazole, 5-Chloro-2-(3,5-di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole, 5-Chloro-2-(3,5-di-tert-butyl-2-hydroxyphenyl)benzotriazole, 5-Chloro-2-(3',5'-di-tert-butyl-2'-hydroxyphenyl)benzotriazole, ADK Stab LA 34, Antioxidant 327, Chiguard 327, Cyasorb UV 5357, Eversorb 75, Hisorb 327, Hisorp 327, Kemisorb 72, LA 34, Light stabilizer 327, Lowilite 27, Mark LA 34, Seesorb 702, Songsorb 3270, TNV 327, Tinuvin 327, UV 2, UV 2 (UV stabilizer), UV 327, UV-Chek AM 607, UVP 327, Viosorb 580
IUPAC Name 2,4-ditert-butyl-6-(5-chlorobenzotriazol-2-yl)phenol
SMILES CC(C)(C)c1cc(c(O)c(c1)C(C)(C)C)n2nc3ccc(Cl)cc3n2
Specifications
Quantity 10 g
InChI Formula InChI=1S/C20H24ClN3O/c1-19(2,3)12-9-14(20(4,5)6)18(25)17(10-12)24-22-15-8-7-13(21)11-16(15)23-24/h7-11,25H,1-6H3
Formula Weight 357.1608
Percent Purity >95%
Chemical Name or Material 2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol
Grade HPLC