missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dichloro-5'-fluoroacetophenone, 97%, Thermo Scientific™

Product Code. 11459677
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11459677

Brand: Thermo Scientific Alfa Aesar B22153.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 704-10-9
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.03
MDL Number MFCD00077499
InChI Key FAKJFAMIABOKBW-UHFFFAOYSA-N
Synonym 2',4'-dichloro-5'-fluoroacetophenone, 1-2,4-dichloro-5-fluorophenyl ethanone, 2,4-dichloro-5-fluoroacetophenone, 1-2,4-dichloro-5-fluorophenyl ethan-1-one, dcfa, acetophenone, 2',4'-dichloro-5'-fluoro, 6-acetyl-1,3-dichloro-4-fluorobenzene, 1-2,4-dichloro-5-fluoro-phenyl ethanone, ethanone, 1-2,4-dichloro-5-fluorophenyl, 1-acetyl-2,4-dichloro-5-fluorobenzene
PubChem CID 727250
IUPAC Name 1-(2,4-dichloro-5-fluorophenyl)ethanone
SMILES CC(=O)C1=CC(F)=C(Cl)C=C1Cl

Specifications

Melting Point 34°C to 36°C
Density 1.425
Boiling Point 134°C to 135°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.546
Quantity 5 g
Beilstein 3090042
Formula Weight 207.03
Percent Purity 97%
Chemical Name or Material 2',4'-Dichloro-5'-fluoroacetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.