missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Dichlorobenzophenone, TRC

Product Code. 30382245
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382245 2.5 g 2.50g
30374380 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382245 Supplier Toronto Research Chemicals Supplier No. TRCD4319752.5G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4'-Dichlorobenzophenone
CAS 85-29-0
Molecular Formula C13 H8 Cl2 O
Quantity 2.5 g
Synonym Benzophenone, 2,4'-dichloro- (6CI,7CI,8CI), (2-Chlorophenyl)(4-chlorophenyl)methanone, 2,4'-Dichlorobenzophenone, NSC 3221, o,p'-DCBP, o,p'-Dichlorobenzophenone
InChI Formula InChI=1S/C13H8Cl2O/c14-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)15/h1-8H
SMILES Clc1ccc(cc1)C(=O)c2ccccc2Cl
IUPAC Name (2-chlorophenyl)-(4-chlorophenyl)methanone
Molecular Weight (g/mol) 251.11
Formula Weight 249.9952
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.