missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoroiodobenzene, 99+%, Thermo Scientific™

Product Code. 10399240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10399240 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10399240 Supplier Thermo Scientific Acros Supplier No. 269780100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2265-93-2
CAS Min % 99.0
CAS Max % 100.0
Color Brown to Yellow
Density 2.0000g/mL
Boiling Point 175°C to 176°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C6H3F2I
Refractive Index 1.5570 to 1.559
MDL Number MFCD00040151
Quantity 10 g
Specific Gravity 2
Synonym 2,4-difluoroiodobenzene, 1,3-difluoro-4-iodobenzene, 2,4-difluoro-1-iodobenze, benzene, 2,4-difluoro-1-iodo, 2,4-difluoro iodobenzene, 2,4-difluoro-1-iodo-benzene, pubchem3463, 2,4-difluoro-iodobenzene, acmc-1cq0v, intermediates-zcf02607
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
InChI Key YKLDMAPEGQYZRT-UHFFFAOYSA-N
SMILES FC1=CC=C(I)C(F)=C1
IUPAC Name 2,4-difluoro-1-iodobenzene
Molecular Weight (g/mol) 239.99
PubChem CID 137519
Formula Weight 239.98
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluoroiodobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.