missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylacetonitrile, 97%, Thermo Scientific™

Product Code. 11478887
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478887 5 g 5g
11468887 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478887 Supplier Thermo Scientific Alfa Aesar Supplier No. B20525.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 656-35-9
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.132
MDL Number MFCD00009971
InChI Key AGAOESUOSOGZOD-UHFFFAOYSA-N
Synonym 2,4-difluorophenylacetonitrile, 2-2,4-difluorophenyl acetonitrile, 2,4-difluorobenzyl cyanide, 2,4-difluorobenzylcyanide, benzeneacetonitrile, 2,4-difluoro, 2,4-difluorophenyl acetonitrile, 2,4-difluoro-phenyl-acetonitrile, 2-2,4-difluorophenyl ethanenitrile, nc1r bf df
PubChem CID 69565
IUPAC Name 2-(2,4-difluorophenyl)acetonitrile
SMILES C1=CC(=C(C=C1F)F)CC#N

Specifications

Density 1.249
Boiling Point 98°C (10 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.48
Quantity 5 g
UN Number UN3276
Beilstein 2614808
Formula Weight 153.13
Percent Purity 97%
Chemical Name or Material 2,4-Difluorophenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.