Learn More
2,4-Dihydroxy-6-methyl-5-nitropyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dihydroxy-6-methyl-5-nitropyrimidine |
| CAS | 16632-21-6 |
| Molecular Formula | C5 H5 N3 O4 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C5H5N3O4/c1-2-3(8(11)12)4(9)7-5(10)6-2/h1H3,(H2,6,7,9,10) |
| SMILES | Cc1nc(O)nc(O)c1[N+](=O)[O-] |
| IUPAC Name | 6-methyl-5-nitropyrimidine-2,4-diol |
| Molecular Weight (g/mol) | 171.11 |
| Formula Weight | 171.028 |