missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxy-Beta-methylcinnamic Acid, TRC

Product Code. 30384023
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384023 100 mg -
30384624 10 mg -
30386884 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384023 Supplier Toronto Research Chemicals Supplier No. TRCD496300100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7706-67-4
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.23
SMILES COc1ccc(\C(=C\C(=O)O)\C)c(OC)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H14O4/c1-8(6-12(13)14)10-5-4-9(15-2)7-11(10)16-3/h4-7H,1-3H3,(H,13,14)/b8-6+
Formula Weight 222.0892
Chemical Name or Material 2,4-Dimethoxy-beta-methylcinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.