missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzyl alcohol, 90%, ACROS Organics™

Product Code. 10211932 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10211932 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10211932 Supplier Thermo Scientific Acros Supplier No. 182810010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16308-92-2
CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Molecular Formula C9H12O
Linear Formula (CH3)2C6H3CH2OH
Refractive Index 1.5330 to 1.5370 (20°C, 589nm)
MDL Number MFCD00004624
Quantity 1 g
Synonym 2,4-dimethylbenzyl alcohol, 2,4-dimethylphenyl methanol, benzenemethanol, 2,4-dimethyl, 2,4-dimethyl-phenyl-methanol, benzyl alcohol, 2,4-dimethyl, 2,4-dimethylbenzylalcohol, acmc-20aoca, benzenemethanol,2,4-dimethyl, 2,4-dimethylphenyl methanol #
InChI Key QUIMJTKRVOBTQN-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)CO)C
IUPAC Name (2,4-dimethylphenyl)methanol
Molecular Weight (g/mol) 136.194
PubChem CID 27809
Formula Weight 136.19
Percent Purity ≥88%
Physical Form Liquid After Melting
Chemical Name or Material 2, 4-Dimethylbenzyl alcohol, 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.