Learn More
2,4-Dimethylphenyl isothiocyanate, 98%, Thermo Scientific™
Product Code. 11302567
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302567 5 g 5g
11312567 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11302567 Supplier Thermo Scientific Alfa Aesar Supplier No. L11665.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39842-01-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00022054
InChI Key HOHSBFCSOARUBF-UHFFFAOYSA-N
Synonym 2,4-dimethylphenyl isothiocyanate, 2,4-dimethylphenylisothiocyanate, 2,4-xylyl isothiocyanate, 1-isothiocyanato-2,4-dimethyl-benzene, 2,4-dimethylbenzenisothiocyanate, xylyl isothiocyanate, 4-isothiocyanato-m-xylene, acmc-1agr7, 2,4-dimethylphenyl-isothiocyanate, 2,4-dimethyl-phenyl isothiocyanate
PubChem CID 142386
IUPAC Name 1-isothiocyanato-2,4-dimethylbenzene
SMILES CC1=CC=C(N=C=S)C(C)=C1
Specifications
Melting Point 25°C to 29°C
Quantity 5 g
Refractive Index 1.625
UN Number UN2923
Beilstein 2690174
Boiling Point 122°C to 130°C (8 mmHg)
Formula Weight 163.24
Percent Purity 98%
Chemical Name or Material 2,4-Dimethylphenyl isothiocyanate