missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylphenyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 11312567
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312567 25 g 25g
11302567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312567 Supplier Thermo Scientific Alfa Aesar Supplier No. L11665.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39842-01-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00022054
InChI Key HOHSBFCSOARUBF-UHFFFAOYSA-N
Synonym 2,4-dimethylphenyl isothiocyanate, 2,4-dimethylphenylisothiocyanate, 2,4-xylyl isothiocyanate, 1-isothiocyanato-2,4-dimethyl-benzene, 2,4-dimethylbenzenisothiocyanate, xylyl isothiocyanate, 4-isothiocyanato-m-xylene, acmc-1agr7, 2,4-dimethylphenyl-isothiocyanate, 2,4-dimethyl-phenyl isothiocyanate
PubChem CID 142386
IUPAC Name 1-isothiocyanato-2,4-dimethylbenzene
SMILES CC1=CC=C(N=C=S)C(C)=C1

Specifications

Melting Point 25°C to 29°C
Quantity 25 g
Refractive Index 1.625
UN Number UN2923
Beilstein 2690174
Boiling Point 122°C to 130°C (8 mmHg)
Formula Weight 163.24
Percent Purity 98%
Chemical Name or Material 2,4-Dimethylphenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.