missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenyl thiocyanate, 98%, Thermo Scientific Chemicals

Product Code. p-7027414
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11411167

Brand: Thermo Scientific Alfa Aesar B24147.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1594-56-5
Molecular Formula C7H3N3O4S
Molecular Weight (g/mol) 225.178
MDL Number MFCD00047702
InChI Key XQDQRCRASHAZBA-UHFFFAOYSA-N
Synonym dntb, nirit, trirodazeen, gryzbol, grzybol, 2,4-dinitrothiocyanobenzene, nirit supra, tri-rodazene, 2,4-dinitrothiocyanatobenzene, rhodandinitrobenzol
PubChem CID 15325
ChEBI CHEBI:53051
IUPAC Name (2,4-dinitrophenyl) thiocyanate
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])SC#N

Specifications

Melting Point 138°C to 140°C
Odor Irritating
Quantity 25 g
Beilstein 1983292
Sensitivity Moisture sensitive
Formula Weight 225.18
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrophenyl thiocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.