missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methanesulfonyl-phenyl)-1-(6-methyl-pyridin-3-yl)-ethanone, TRC

Product Code. 17942705
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17942705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17942705 Supplier Toronto Research Chemicals Supplier No. TRCM2259151G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 221615-75-4
Molecular Formula C15 H15 N O3 S
Quantity 1 g
Synonym 1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]-ethanone, 1-(6-Methylpyridin-3-yl)-2-[4-(methylsulfonyl)phenyl]ethanone
InChI Formula InChI=1S/C15H15NO3S/c1-11-3-6-13(10-16-11)15(17)9-12-4-7-14(8-5-12)20(2,18)19/h3-8,10H,9H2,1-2H3
SMILES Cc1ccc(cn1)C(=O)Cc2ccc(cc2)S(=O)(=O)C
IUPAC Name 1-(6-methylpyridin-3-yl)-2-(4-methylsulfonylphenyl)ethanone
Molecular Weight (g/mol) 289.35
Formula Weight 289.0773
Percent Purity >95%
Chemical Name or Material 1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]ethanone
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.