missing translation for 'onlineSavingsMsg'
Learn More

2-[4-[(Oxiran-2-yl)methoxy]phenyl]acetonitrile, Mikromol

Product Code. 30457423
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457423 4 x 25 mg -
17930986 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30457423 Supplier Mikromol Supplier No. MM0013.0900254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[4-[(Oxiran-2-yl)methoxy]phenyl]acetonitrile, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(2,3-Epoxypropoxy)phenylacetonitrile
CAS 35198-42-6
Analyte or Component Names 4-(2,3-Epoxypropoxy)phenylacetonitrile
Shipping Condition Ice pack (-20°C)
Molecular Formula C11 H11 N O2
Certifications/Compliance ISO 9001
InChI Formula InChI=1S/C11H11NO2/c12-6-5-9-1-3-10(4-2-9)13-7-11-8-14-11/h1-4,11H,5,7-8H2
Solution Type Neat
SMILES N#CCc1ccc(OCC2CO2)cc1
IUPAC Name 2-[4-(oxiran-2-ylmethoxy)phenyl]acetonitrile
Molecular Weight (g/mol) 189.21
Quantity 4 x 25 mg
Formula Weight 189.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.