missing translation for 'onlineSavingsMsg'
Learn More

2-(4-phenyl-1,3-thiazol-2-yl)acetamide, TRC

Product Code. 30423896
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423896 50 mg -
30419929 100 mg -
30410623 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423896 Supplier Toronto Research Chemicals Supplier No. TRCP32288550MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58351-19-2
Molecular Formula C11H10N2OS
Molecular Weight (g/mol) 218.27
IUPAC Name 2-(4-phenyl-1,3-thiazol-2-yl)acetamide
SMILES NC(=O)Cc1nc(cs1)c2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H10N2OS/c12-10(14)6-11-13-9(7-15-11)8-4-2-1-3-5-8/h1-5,7H,6H2,(H2,12,14)
Formula Weight 218.0514
Chemical Name or Material 2-(4-phenyl-1,3-thiazol-2-yl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.