Learn More
2-(4-Pyridyl)benzimidazole, 97%
Product Code. 15498227
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498227 25 g 25g
15488227 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15498227 Supplier Thermo Scientific Alfa Aesar Supplier No. H51778.14
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
CAS: 2208-59-5 | C12H9N3 | 195.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2208-59-5
Molecular Formula C12H9N3
Molecular Weight (g/mol) 195.23
MDL Number MFCD00453948
InChI Key UYWWLYCGNNCLKE-UHFFFAOYSA-N
Synonym 2-4-pyridyl benzimidazole, 2-pyridin-4-yl-1h-benzo d imidazole, 2-pyridin-4-yl-1h-benzimidazole, 2-pyridin-4-yl-1h-1,3-benzodiazole, 2-4-pyridyl-1h-benzimidazole, 2-pyridin-4-yl-1h-benzoimidazole, 2-benzimidazole, acmc-1ch3h, cambridge id 5231114, 2-4-pyridinyl-benzimidazole
PubChem CID 247634
IUPAC Name 2-pyridin-4-yl-1H-benzimidazole
SMILES N1C2=CC=CC=C2N=C1C1=CC=NC=C1
Specifications
Melting Point 216°C to 221°C
Odor Odorless
Quantity 25 g
Formula Weight 195.22
Percent Purity 97%
Chemical Name or Material 2-(4-Pyridyl)benzimidazole