Learn More
2,5-Diamino-4,6-dichloropyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 4,6-Dichloropyrimidine-2,5-diamine |
| CAS | 55583-59-0 |
| Molecular Formula | C4 H4 Cl2 N4 |
| Quantity | 250 mg |
| Synonym | 2,5-Pyrimidinediamine, 4,6-dichloro-, 4,6-Dichloro-2,5-pyrimidinediamine, 2,5-Diamino-4,6-dichloropyrimidine, 4,6-Dichloropyrimidine-2,5-diamine, NSC 176971 |
| InChI Formula | InChI=1S/C4H4Cl2N4/c5-2-1(7)3(6)10-4(8)9-2/h7H2,(H2,8,9,10) |
| SMILES | Nc1nc(Cl)c(N)c(Cl)n1 |
| IUPAC Name | 4,6-dichloropyrimidine-2,5-diamine |
| Molecular Weight (g/mol) | 179.01 |
| Formula Weight | 177.9813 |