Learn More
2,5-Dichloro-p-xylene, 98%
Product Code. 11489267
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489267 25 g 25g
11499267 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11489267 Supplier Thermo Scientific Alfa Aesar Supplier No. B21344.14
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
CAS: 1124-05-6 | C8H8Cl2 | 175.052 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1124-05-6
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000610
InChI Key UTGSRNVBAFCOEU-UHFFFAOYSA-N
Synonym 2,5-dichloro-p-xylene, 2,5-dichloro-1,4-dimethylbenzene, benzene, 1,4-dichloro-2,5-dimethyl, p-xylene, 2,5-dichloro, unii-7p8m86zob2, 2,5-dichloro-para-xylene, p-xylene,5-dichloro, p-xylene,2,5-dichloro, acmc-2099eq, dsstox_cid_24780
PubChem CID 70755
IUPAC Name 1,4-dichloro-2,5-dimethylbenzene
SMILES CC1=CC(=C(C=C1Cl)C)Cl
Specifications
Melting Point 68°C to 70°C
Quantity 25 g
Boiling Point 221°C to 223°C
Formula Weight 175.06
Percent Purity 98%
Chemical Name or Material 2,5-Dichloro-p-xylene