missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichlorobiphenyl-2',3',4',5',6'-d5, CDN
Click to view available options
Quantity:
0.01 g
Specifications
Specifications
| Chemical Name or Material | 2,5-Dichlorobiphenyl-2',3',4',5',6'-D5 |
| CAS | 1219804-50-8 |
| Molecular Formula | C12 D5 H3 Cl2 |
| Quantity | 0.01 g |
| Synonym | 2,5-Dichloro-1,1'-biphenyl-2',3',4',5',6'-d5, 2,5-Dichlorobiphenyl-2',3',4',5',6'-d5, 2,5-Dichlorobiphenyl-2',3',4',5',6'-D5, PCB 9-d5, 2,5-DiCB-2',3',4',5',6'-d5 |
| InChI Formula | InChI=1 S/C12H8Cl2/c13-10-6-7-12(14)11(8-10)9-4-2-1-3-5-9/h1-8 H/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2cc(Cl)ccc2Cl |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(2,5-dichlorophenyl)benzene |
| Molecular Weight (g/mol) | 228.1287 |
| Formula Weight | 227.0317 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction