missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroisonicotinic acid, 97%, Thermo Scientific™

Product Code. 11996441 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11996441

Brand: Thermo Scientific Acros 442830010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 88912-26-9
Melting Point 227°C to 229°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C6H3Cl2NO2
Quantity 1 g
Synonym 2,5-dichloroisonicotinic acid, 2,5-dichloro-isonicotinic acid, 4-carboxy-2,5-dichloropyridine, 4-pyridinecarboxylic acid, 2,5-dichloro, 2,5-dichloro-4-pyridinecarboxylic acid, pubchem13559, acmc-209qxv, ksc447s3n, 2,5-dichloro isonicotinic acid, gfovttqvbdeypp-uhfffaoysa
InChI Key GFOVTTQVBDEYPP-UHFFFAOYSA-N
SMILES C1=C(C(=CN=C1Cl)Cl)C(=O)O
IUPAC Name 2,5-dichloropyridine-4-carboxylic acid
Molecular Weight (g/mol) 192
PubChem CID 2736087
Formula Weight 192
Percent Purity 97%
Chemical Name or Material 2, 5-Dichloroisonicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dichloroisonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.