Learn More
2,5-Dichlorothiobenzamide, 97%, Thermo Scientific™
Product Code. 15485037
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15485037 250 mg 250mg
15475037 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15485037 Supplier Thermo Scientific Alfa Aesar Supplier No. H32323.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69622-81-7
Molecular Formula C7H5Cl2NS
Molecular Weight (g/mol) 206.084
MDL Number MFCD09025686
InChI Key YQORNGRDGWVQOD-UHFFFAOYSA-N
Synonym 2,5-dichlorothiobenzamide, benzenecarbothioamide,2,5-dichloro, 2,5-dichlorobenzene-1-carbothioamide
PubChem CID 45040719
IUPAC Name 2,5-dichlorobenzenecarbothioamide
SMILES C1=CC(=C(C=C1Cl)C(=S)N)Cl
Specifications
Melting Point 88°C to 92°C
Quantity 250 mg
UN Number UN2811
Formula Weight 206.09
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorothiobenzamide